Assay Details
Gene Symbol: Maf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164607.1 | NP_001158079.1 | 8 - 9 | 1220 | 66 | |
NM_001164608.1 | NP_001158080.1 | 7 - 8 | 1127 | 66 | ||
NM_026859.3 | NP_081135.3 | 7 - 8 | 1174 | 66 | ||
GenBank mRNA | AK004406.1 | - | 7 - 8 | 847 | 66 | |
AK042913.1 | - | 7 - 8 | 1068 | 66 | ||
AK154047.1 | - | 7 - 8 | 1117 | 66 | ||
AK189411.1 | - | 1 - 2 | 48 | 66 | ||
BB250147.2 | - | 1 - 2 | 132 | 66 | ||
BC016260.1 | - | 7 - 8 | 1162 | 4914167 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transcription from RNA polymerase III promoter
transcription factor TFIIIB complex assembly
RNA polymerase III core binding
RNA polymerase III type 1 promoter DNA binding
RNA polymerase III type 2 promoter DNA binding
RNA polymerase III type 3 promoter DNA binding
GABA receptor binding
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transcription from RNA polymerase III promoter
transcription factor TFIIIB complex assembly
RNA polymerase III core binding
RNA polymerase III type 1 promoter DNA binding
RNA polymerase III type 2 promoter DNA binding
RNA polymerase III type 3 promoter DNA binding
GABA receptor binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347742 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347741 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347743 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347744 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406365 | BC016260.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406365 | AK154047.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406365 | AK042913.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406365 | AK214780.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406365 | BB250147.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406365 | BC016260.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406365 | AK154047.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406365 | AK042913.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406365 | AK214780.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406365 | BB250147.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79551 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 58176 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406365 | BC016260.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406365 | AK154047.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406365 | AK042913.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406365 | AK214780.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406365 | BB250147.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406365 | BC016260.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406365 | AK154047.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406365 | AK042913.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406365 | AK214780.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406365 | BB250147.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347773 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347775 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347780 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347749 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347784 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347769 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347767 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347779 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347763 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347756 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347750 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347770 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347768 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347771 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347762 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347785 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347782 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347742 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347751 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347766 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347743 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347776 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347759 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08422019 | - | - |
Whole Mouse Genome G4122A | A_51_P406365 | BC016260.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406365 | AK154047.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406365 | AK042913.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406365 | AK214780.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406365 | BB250147.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406365 | BC016260.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406365 | AK154047.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406365 | AK042913.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406365 | AK214780.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406365 | BB250147.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108504 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12345065 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347743 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347742 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347744 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347751 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347771 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347790 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12345041 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347754 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347769 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347766 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347779 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347763 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347805 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12345026 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12345078 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347743 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347793 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347791 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347762 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347800 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347796 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347787 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12345035 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347797 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12345059 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347807 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12345064 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347750 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12345043 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12345081 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347764 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347746 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347759 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347756 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12345117 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347768 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30347748 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12345118 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10424837 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10424841 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10424839 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10424835 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10424834 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10424840 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10424836 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10424838 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1420047_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451126_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1420046_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10424833 | - | - |
Mouse Genome 430 2.0 Array | 1451126_at | - | - |
Mouse Genome 430 2.0 Array | 1420046_s_at | - | - |
Mouse Genome 430 2.0 Array | 1451126_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1420046_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1420047_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1420047_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68877_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68877_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1500004126.mm.1 | NM_001164607.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004126.mm.1 | NM_001164607.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004126.mm.1 | NM_001164608.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004126.mm.1 | NM_001164608.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004126.mm.1 | NM_026859.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004126.mm.1 | NM_026859.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004126.mm.1 | TC1500000588.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500004127.mm.1 | NM_001164607.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004127.mm.1 | NM_001164607.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004127.mm.1 | NM_001164608.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004127.mm.1 | NM_001164608.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004127.mm.1 | NM_026859.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004127.mm.1 | NM_026859.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004127.mm.1 | TC1500000588.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500004124.mm.1 | NM_001164607.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004124.mm.1 | NM_001164607.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004124.mm.1 | NM_001164608.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004124.mm.1 | NM_001164608.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004124.mm.1 | NM_026859.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004124.mm.1 | NM_026859.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004124.mm.1 | TC1500000588.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500004125.mm.1 | NM_001164607.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004125.mm.1 | NM_001164607.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004125.mm.1 | NM_001164608.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004125.mm.1 | NM_001164608.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004125.mm.1 | NM_026859.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004125.mm.1 | NM_026859.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004125.mm.1 | TC1500000588.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500004123.mm.1 | NM_001164607.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004123.mm.1 | NM_001164607.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004123.mm.1 | NM_001164608.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004123.mm.1 | NM_001164608.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004123.mm.1 | NM_026859.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004123.mm.1 | NM_026859.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500004123.mm.1 | TC1500000588.mm.1 | MTA TC |
Mouse Expression Set 430 | 1451126_at | - | - |
Mouse Expression Set 430 | 1451126_PM_at | - | - |
Murine Genome U74v2 Set | 93490_at | - | - |
Murine Genome U74v2 Set | 93490_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451126_at | - | - |
Mouse Genome 430A 2.0 Array | 1451126_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451126_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420047_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420047_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420046_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451126_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420046_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 520524 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000520524 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2741537 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2741537 | - | - |
mouse-6 v1.1 (Illumina) | scl068877.8_322-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 520524 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl068877.8_322-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 520524 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2741538 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2741538 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl068877.8_322-S | - | - |
