Assay Details
| Gene Symbol: Maf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164607.1 | NP_001158079.1 | 2 - 3 | 415 | 80 | |
| GenBank mRNA | CK619143.1 | - | 2 - 3 | 360 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001164607.1 transcripts.
Set Membership: |
|

Maf1
Genomic Map