Assay Details
Gene Symbol: Sharpin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025340.2 | NP_079616.2 | 3 - 4 | 939 | 94 | |
GenBank mRNA | AB052763.1 | - | 3 - 4 | 925 | 94 | |
AK002878.1 | - | 3 - 4 | 510 | 94 | ||
AK003578.1 | - | 3 - 4 | 520 | 93 | ||
AK165824.1 | - | 2 - 3 | 719 | 94 | ||
BC016203.1 | - | 3 - 4 | 880 | 4527839 | 94 | |
BC055758.1 | - | 3 - 4 | 939 | 6415354 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
