Assay Details
| Gene Symbol: Stambpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029682.4 | NP_083958.3 | 8 - 9 | 1454 | 66 | |
| GenBank mRNA | AB010121.1 | - | 8 - 9 | 1149 | 66 | |
| AB010122.1 | - | 8 - 9 | 1113 | 66 | ||
| AB066211.1 | - | 8 - 9 | 1454 | 66 | ||
| AB066212.1 | - | 4 - 5 | 803 | 66 | ||
| AB066213.1 | - | 8 - 9 | 1137 | 66 | ||
| AK007087.1 | - | 4 - 5 | 787 | 66 | ||
| BC018343.1 | - | 8 - 9 | 1180 | 4010423 | 66 | |
| BC023097.1 | - | 8 - 9 | 1423 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stambpl1
Genomic Map