Assay Details
| Gene Symbol: Stambpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029682.4 | NP_083958.3 | 9 - 10 | 1564 | 57 | |
| GenBank mRNA | AB010121.1 | - | 9 - 10 | 1259 | 57 | |
| AB010122.1 | - | 9 - 10 | 1223 | 57 | ||
| AB066211.1 | - | 9 - 10 | 1564 | 57 | ||
| AB066212.1 | - | 5 - 6 | 913 | 57 | ||
| AB066213.1 | - | 9 - 10 | 1247 | 57 | ||
| AK007087.1 | - | 5 - 6 | 897 | 57 | ||
| BC018343.1 | - | 9 - 10 | 1290 | 4010423 | 57 | |
| BC023097.1 | - | 9 - 10 | 1533 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stambpl1
Genomic Map