Assay Details
| Gene Symbol: Dtx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256097.1 | NP_001243026.1 | 6 - 7 | 1498 | 98 | |
| NM_001256098.1 | NP_001243027.1 | 5 - 6 | 1420 | 98 | ||
| GenBank mRNA | AB015424.1 | - | 5 - 6 | 1343 | 98 | |
| AK044105.1 | - | 6 - 7 | 1455 | 98 | ||
| AK054443.1 | - | 5 - 6 | 1410 | 98 | ||
More Information
Set Membership: |
|

Dtx2
Genomic Map