Assay Details
| Gene Symbol: Pde6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008806.2 | NP_032832.2 | 16 - 17 | 2088 | 149 | |
| GenBank mRNA | AK039397.1 | - | 16 - 17 | 1800 | 149 | |
| AK044364.1 | - | 16 - 17 | 2031 | 149 | ||
| BC048247.1 | - | 16 - 17 | 2067 | 149 | ||
| BC058803.1 | - | 14 - 15 | 2947 | 149 | ||
| BC129923.1 | - | 16 - 17 | 2018 | 40111882 | 149 | |
| BC129924.1 | - | 16 - 17 | 2017 | 149 | ||
| X55968.1 | - | 16 - 17 | 2021 | 149 | ||
| X60133.1 | - | 16 - 17 | 2028 | 149 | ||
| X87952.1 | - | 16 - 17 | 2028 | 149 | ||
More Information
Set Membership: |
|

Pde6b
Genomic Map