Assay Details
| Gene Symbol: Cd37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290802.1 | NP_001277731.1 | 6 - 7 | 612 | 65 | |
| NM_001290804.1 | NP_001277733.1 | 5 - 6 | 537 | 65 | ||
| NM_007645.4 | NP_031671.1 | 5 - 6 | 561 | 65 | ||
| GenBank mRNA | AK154466.1 | - | 6 - 7 | 582 | 65 | |
| AK155663.1 | - | 5 - 6 | 538 | 65 | ||
| AK155966.1 | - | 5 - 6 | 526 | 65 | ||
| BC019402.1 | - | 5 - 6 | 598 | 4013070 | 65 | |
| BE849269.1 | - | 5 - 6 | 479 | 65 | ||
| CT010217.1 | - | 5 - 6 | 450 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd37
Genomic Map