| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500013610.mm.1 |
TC0500001508.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500013610.mm.1 |
NM_025450.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013610.mm.1 |
NM_025450.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013612.mm.1 |
TC0500001508.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500013612.mm.1 |
NM_025450.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013612.mm.1 |
NM_025450.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013617.mm.1 |
TC0500001508.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500013617.mm.1 |
NM_025450.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013617.mm.1 |
NM_025450.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013611.mm.1 |
TC0500001508.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500013611.mm.1 |
NM_025450.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013611.mm.1 |
NM_025450.4 |
Refseq |