Assay Details
| Gene Symbol: Pop5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026398.4 | NP_080674.1 | 1 - 2 | 35 | 64 | |
| GenBank mRNA | BC022670.1 | - | 1 - 2 | 59 | 4224973 | 64 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC022670.1, NM_026398.4 transcripts.
Set Membership: |
|

Pop5
Genomic Map