Assay Details
| Gene Symbol: Daglb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144915.3 | NP_659164.2 | 2 - 3 | 437 | 57 | |
| GenBank mRNA | AK086747.1 | - | 2 - 3 | 347 | 57 | |
| AK087884.1 | - | 2 - 3 | 292 | 57 | ||
| AK135554.1 | - | 2 - 3 | 317 | 57 | ||
| AK136372.1 | - | 2 - 3 | 295 | 57 | ||
| BC016105.1 | - | 2 - 3 | 333 | 4921222 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK086747.1 transcripts.
Set Membership: |
|

Daglb
Genomic Map