Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500008417.mm.1 |
TC0500000997.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008417.mm.1 |
NM_029270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008417.mm.1 |
NM_029270.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008417.mm.1 |
NM_146161.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008417.mm.1 |
NM_146161.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008415.mm.1 |
TC0500000997.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008415.mm.1 |
NM_029270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008415.mm.1 |
NM_029270.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008415.mm.1 |
NM_146161.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008415.mm.1 |
NM_146161.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008419.mm.1 |
TC0500000997.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008419.mm.1 |
NM_029270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008419.mm.1 |
NM_029270.2 |
Refseq |