Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1440213_PM_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1440213_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1440213_PM_a_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0500030576.mm.1 |
TC0500003387.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030576.mm.1 |
NR_026688.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030576.mm.1 |
NR_026688.1 |
Refseq |
Mouse Expression Set 430 |
1440213_PM_a_at |
- |
- |
Mouse Expression Set 430 |
1440213_a_at |
- |
- |
Murine Genome U74v2 Set |
103355_PM_at |
- |
- |
Murine Genome U74v2 Set |
103355_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1440213_a_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1440213_PM_a_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1440213_PM_a_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1440213_a_at |
- |
- |