Assay Details
| Gene Symbol: Srrm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026886.3 | NP_081162.1 | 5 - 6 | 815 | 71 | |
| GenBank mRNA | AK053836.1 | - | 5 - 6 | 547 | 71 | |
| AK165719.1 | - | 5 - 6 | 533 | 71 | ||
| BC056392.1 | - | 5 - 6 | 805 | 6855144 | 71 | |
| JN860221.1 | - | 5 - 6 | 462 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srrm4
Genomic Map