Assay Details
Gene Symbol: Prkab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031869.2 | NP_114075.1 | 2 - 3 | 506 | 67 | |
GenBank mRNA | AK088250.1 | - | 2 - 3 | 469 | 67 | |
AK153552.1 | - | 2 - 3 | 526 | 67 | ||
BC016398.1 | - | 2 - 3 | 506 | 3585002 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
