Assay Details
| Gene Symbol: Svop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026805.1 | NP_081081.1 | 11 - 12 | 1236 | 72 | |
| GenBank mRNA | AK029670.1 | - | 10 - 11 | 945 | 72 | |
| AK047655.1 | - | 11 - 12 | 1229 | 72 | ||
| AK047764.1 | - | 11 - 12 | 1237 | 72 | ||
| AK053833.1 | - | 11 - 12 | 1227 | 72 | ||
| AK082744.1 | - | 11 - 12 | 1238 | 72 | ||
| BC059878.1 | - | 11 - 12 | 1189 | 6812082 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Svop
Genomic Map