Assay Details
| Gene Symbol: Svop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026805.1 | NP_081081.1 | 13 - 14 | 1460 | 90 | |
| GenBank mRNA | AK029670.1 | - | 12 - 13 | 1169 | 90 | |
| AK047655.1 | - | 13 - 14 | 1453 | 90 | ||
| AK047764.1 | - | 13 - 14 | 1461 | 90 | ||
| AK053833.1 | - | 13 - 14 | 1451 | 90 | ||
| AK082744.1 | - | 13 - 14 | 1462 | 90 | ||
| BC059878.1 | - | 13 - 14 | 1413 | 6812082 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Svop
Genomic Map