Assay Details
Gene Symbol: Lfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008494.3 | NP_032520.1 | 5 - 6 | 893 | 85 | |
GenBank mRNA | AF015768.1 | - | 5 - 6 | 922 | 85 | |
AK004642.1 | - | 5 - 6 | 894 | 85 | ||
AK153283.1 | - | 5 - 6 | 894 | 85 | ||
BC115680.1 | - | 4 - 5 | 559 | 40104178 | 85 | |
BC138600.1 | - | 5 - 6 | 914 | 8861620 | 85 | |
BC138601.1 | - | 5 - 6 | 914 | 8861621 | 85 | |
U94351.1 | - | 5 - 6 | 858 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
