Assay Details
| Gene Symbol: Tmem120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172541.2 | NP_766129.1 | 7 - 8 | 668 | 88 | |
| GenBank mRNA | AK028119.1 | - | 7 - 8 | 669 | 88 | |
| AK050367.1 | - | 2 - 3 | 158 | 88 | ||
| BC046757.1 | - | 7 - 8 | 628 | 5693443 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem120a
Genomic Map