Assay Details
| Gene Symbol: Tmem120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172541.2 | NP_766129.1 | 8 - 9 | 731 | 80 | |
| GenBank mRNA | AK028119.1 | - | 8 - 9 | 732 | 80 | |
| AK050367.1 | - | 3 - 4 | 221 | 80 | ||
| BC046757.1 | - | 8 - 9 | 691 | 5693443 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem120a
Genomic Map