Assay Details
| Gene Symbol: Adap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172723.4 | NP_766311.2 | 7 - 8 | 841 | 63 | |
| GenBank mRNA | AK076754.1 | - | 6 - 7 | 827 | 63 | |
| AK076768.1 | - | 7 - 8 | 598 | 63 | ||
| BC056934.1 | - | 6 - 7 | 567 | 63 | ||
| BC079634.1 | - | 6 - 7 | 635 | 63 | ||
| BC144874.1 | - | 6 - 7 | 586 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adap1
Genomic Map