Assay Details
Gene Symbol: Tyw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015876.2 | NP_001015876.1 | 2 - 3 | 280 | 76 | |
NM_178897.4 | NP_849228.1 | 2 - 3 | 150 | 76 | ||
GenBank mRNA | AK045279.1 | - | 2 - 3 | 151 | 76 | |
AK082884.1 | - | 2 - 3 | 271 | 76 | ||
AK162899.1 | - | 2 - 3 | 283 | 76 | ||
BC068126.1 | - | 2 - 3 | 188 | 6829456 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
