Assay Details
| Gene Symbol: Tyw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015876.2 | NP_001015876.1 | 5 - 6 | 704 | 59 | |
| NM_178897.4 | NP_849228.1 | 5 - 6 | 574 | 59 | ||
| GenBank mRNA | AK045279.1 | - | 5 - 6 | 575 | 59 | |
| AK082884.1 | - | 5 - 6 | 695 | 59 | ||
| AK162899.1 | - | 5 - 6 | 707 | 59 | ||
| BC068126.1 | - | 5 - 6 | 612 | 6829456 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tyw1
Genomic Map