Assay Details
| Gene Symbol: 1700028K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175241.1 | NP_780450.1 | 4 - 5 | 457 | 93 | |
| NM_182745.2 | NP_877422.1 | 4 - 5 | 457 | 93 | ||
| GenBank mRNA | AK006463.1 | - | 4 - 5 | 457 | 93 | |
| AK032886.1 | - | 5 - 6 | 657 | 93 | ||
| CN835533.1 | - | 4 - 5 | 432 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

1700028K03Rik
Genomic Map