Assay Details
| Gene Symbol: Naa25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172722.3 | NP_766310.2 | 5 - 6 | 152 | ||
| GenBank mRNA | AK049325.1 | - | 5 - 6 | 152 | ||
| AK052060.1 | - | 5 - 6 | 152 | |||
| AK149758.1 | - | 5 - 6 | 152 | |||
| AK165175.1 | - | 5 - 6 | 152 | |||
| AK167405.1 | - | 5 - 6 | 152 | |||
| AK167491.1 | - | 5 - 6 | 152 | |||
| BC028868.1 | - | 3 - 4 | 152 | |||
| BC037595.1 | - | 5 - 6 | 472 | 152 | ||
| BC055480.1 | - | 4 - 5 | 152 | |||
| BC082314.1 | - | 5 - 6 | 152 | |||
| BC106182.1 | - | 5 - 6 | 152 | |||
| BC132172.1 | - | 5 - 6 | 40130449 | 152 | ||
| BC138093.1 | - | 5 - 6 | 8861112 | 152 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Naa25
Genomic Map