Assay Details
Gene Symbol: Usp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029749.2 | NP_084025.2 | 6 - 7 | 843 | 70 | |
GenBank mRNA | AK010801.1 | - | 5 - 6 | 653 | 70 | |
AY336567.1 | - | 5 - 6 | 716 | 70 | ||
BC137852.1 | - | 5 - 6 | 736 | 8860871 | 70 | |
BG079818.2 | - | 2 - 3 | 125 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
