Assay Details
| Gene Symbol: Eif2ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013557.2 | NP_038585.2 | 1326 | 59 | ||
| GenBank mRNA | AK156492.1 | - | 1285 | 59 | ||
| AK173160.1 | - | 1276 | 59 | |||
| AY033898.1 | - | 1311 | 59 | |||
| BC028923.1 | - | 1298 | 4164417 | 59 | ||
| BC111035.1 | - | 1277 | 5357121 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif2ak1
Genomic Map