Assay Details
| Gene Symbol: Eif2ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013557.2 | NP_038585.2 | 1424 | 100 | ||
| GenBank mRNA | AK156492.1 | - | 1383 | 100 | ||
| AK173160.1 | - | 1374 | 100 | |||
| AK184294.1 | - | 2 - 3 | 125 | 100 | ||
| AY033898.1 | - | 1409 | 100 | |||
| BC028923.1 | - | 1396 | 4164417 | 100 | ||
| BC111035.1 | - | 1375 | 5357121 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif2ak1
Genomic Map