Assay Details
| Gene Symbol: Eif2ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013557.2 | NP_038585.2 | 380 | 63 | ||
| GenBank mRNA | AK014775.1 | - | 342 | 63 | ||
| AK020887.1 | - | 226 | 63 | |||
| AK155206.1 | - | 362 | 63 | |||
| AK156492.1 | - | 341 | 63 | |||
| AK158437.1 | - | 336 | 63 | |||
| AK173160.1 | - | 330 | 63 | |||
| AY033898.1 | - | 365 | 63 | |||
| BB625879.1 | - | 381 | 63 | |||
| BC028923.1 | - | 352 | 4164417 | 63 | ||
| BC111035.1 | - | 331 | 5357121 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif2ak1
Genomic Map