Assay Details
| Gene Symbol: Eif2ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013557.2 | NP_038585.2 | 550 | 76 | ||
| GenBank mRNA | AK014775.1 | - | 512 | 76 | ||
| AK155206.1 | - | 532 | 76 | |||
| AK156492.1 | - | 511 | 76 | |||
| AK158437.1 | - | 506 | 76 | |||
| AK173160.1 | - | 500 | 76 | |||
| AY033898.1 | - | 535 | 76 | |||
| BB625879.1 | - | 551 | 76 | |||
| BC028923.1 | - | 522 | 4164417 | 76 | ||
| BC111035.1 | - | 501 | 5357121 | 76 | ||
More Information
Set Membership: |
|

Eif2ak1
Genomic Map