Assay Details
| Gene Symbol: Eif2ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013557.2 | NP_038585.2 | 646 | 129 | ||
| GenBank mRNA | AK155206.1 | - | 628 | 129 | ||
| AK158437.1 | - | 602 | 129 | |||
| AK173160.1 | - | 596 | 129 | |||
| AY033898.1 | - | 631 | 129 | |||
| BC028923.1 | - | 618 | 4164417 | 129 | ||
| BC111035.1 | - | 597 | 5357121 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif2ak1
Genomic Map