Assay Details
| Gene Symbol: Rps5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009095.2 | NP_033121.2 | 1 - 2 | 72 | 106 | |
| GenBank mRNA | AK002235.1 | - | 1 - 2 | 66 | 106 | |
| AK009663.1 | - | 1 - 2 | 66 | 106 | ||
| AK010681.1 | - | 1 - 2 | 66 | 106 | ||
| AK012424.1 | - | 1 - 2 | 66 | 106 | ||
| AK012481.1 | - | 1 - 2 | 66 | 106 | ||
| AK020477.1 | - | 1 - 2 | 65 | 106 | ||
| AK020754.1 | - | 1 - 2 | 66 | 106 | ||
| AK050611.1 | - | 1 - 2 | 66 | 106 | ||
| AK050622.1 | - | 1 - 2 | 66 | 106 | ||
| AK152449.1 | - | 1 - 2 | 66 | 106 | ||
| AK160552.1 | - | 1 - 2 | 66 | 106 | ||
| AK164079.1 | - | 1 - 2 | 66 | 106 | ||
| AK167533.1 | - | 1 - 2 | 66 | 106 | ||
| AK168044.1 | - | 1 - 2 | 66 | 106 | ||
| AK168106.1 | - | 1 - 2 | 66 | 106 | ||
| AK169016.1 | - | 1 - 2 | 66 | 106 | ||
| AK172446.1 | - | 1 - 2 | 66 | 106 | ||
| BC020401.1 | - | 1 - 2 | 46 | 106 | ||
| BC058690.1 | - | 1 - 2 | 53 | 6814814 | 106 | |
| BY024242.1 | - | 1 - 2 | 72 | 106 | ||
| U78085.1 | - | 1 - 2 | 58 | 106 | ||
| Y12431.1 | - | 1 - 2 | 48 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002235.1, AK009663.1, AK010681.1, AK012424.1, AK012481.1, AK020477.1, AK020754.1, AK050611.1, AK050622.1, AK152449.1, AK160552.1, AK164079.1, AK167533.1, AK168044.1, AK168106.1, AK169016.1, AK172446.1, BC058690.1, NM_009095.2, U78085.1, Y12431.1 transcripts.
Set Membership: |
|

Rps5
Genomic Map