Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700027640.mm.1 |
NM_001163733.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027640.mm.1 |
NM_001163733.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027640.mm.1 |
NM_023649.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027640.mm.1 |
NM_023649.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027640.mm.1 |
NM_153677.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027640.mm.1 |
NM_153677.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027640.mm.1 |
TC0700003032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027641.mm.1 |
NM_001163733.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027641.mm.1 |
NM_001163733.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027641.mm.1 |
NM_023649.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027641.mm.1 |
NM_023649.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027641.mm.1 |
NM_153677.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027641.mm.1 |
NM_153677.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027641.mm.1 |
TC0700003032.mm.1 |
MTA TC |