Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700007146.mm.1 |
TC0700000834.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007146.mm.1 |
NM_053256.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007146.mm.1 |
NM_053256.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007145.mm.1 |
TC0700000834.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007145.mm.1 |
NM_053256.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007145.mm.1 |
NM_053256.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007141.mm.1 |
TC0700000834.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007141.mm.1 |
NM_053256.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007141.mm.1 |
NM_053256.2 |
Refseq |