Assay Details
| Gene Symbol: Car11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009800.4 | NP_033930.1 | 6 - 7 | 949 | 63 | |
| GenBank mRNA | AB079601.1 | - | 6 - 7 | 728 | 63 | |
| AF050105.1 | - | 4 - 5 | 502 | 63 | ||
| AK013465.1 | - | 6 - 7 | 949 | 63 | ||
| AK140073.1 | - | 6 - 7 | 1050 | 63 | ||
| AY075026.1 | - | 6 - 7 | 950 | 63 | ||
| BC019393.1 | - | 6 - 7 | 934 | 3968862 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car11
Genomic Map