Assay Details
| Gene Symbol: Car11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009800.4 | NP_033930.1 | 8 - 9 | 1269 | 79 | |
| GenBank mRNA | AB079601.1 | - | 8 - 9 | 1048 | 79 | |
| AK013465.1 | - | 8 - 9 | 1269 | 79 | ||
| AK140073.1 | - | 8 - 9 | 1370 | 79 | ||
| AY075026.1 | - | 8 - 9 | 1270 | 79 | ||
| BC019393.1 | - | 8 - 9 | 1254 | 3968862 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25094544 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25094545 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25094546 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480013 | AY075026.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480013 | BC019393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480013 | NM_009800.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480013 | AB079601.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480013 | AF050105.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480013 | AY075026.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480013 | BC019393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480013 | NM_009800.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480013 | AB079601.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480013 | AF050105.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150691 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29791 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480013 | AY075026.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480013 | BC019393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480013 | NM_009800.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480013 | AB079601.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480013 | AF050105.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480013 | AY075026.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480013 | BC019393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480013 | NM_009800.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480013 | AB079601.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480013 | AF050105.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25094552 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06183732 | - | - |
| Whole Mouse Genome G4122A | A_51_P480013 | AY075026.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480013 | BC019393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480013 | NM_009800.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480013 | AB079601.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480013 | AF050105.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480013 | AY075026.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480013 | BC019393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480013 | NM_009800.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480013 | AB079601.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480013 | AF050105.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110215 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139354 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06165348 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06165347 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06323327 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06122755 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06122750 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06323331 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25094544 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25094546 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25094545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25094556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06323329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06323322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06165342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06122753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06612341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06122742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06122750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06047067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06165352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06165337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06323328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06122752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06165345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06402666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06323327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06612345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25094520 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10553091 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553084 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553090 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553086 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553085 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553087 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553081 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553088 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553089 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553083 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424379_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10553080 | - | - |
| Mouse Genome 430 2.0 Array | 1424379_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424379_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12348_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12348_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700007554.mm.1 | TC0700000860.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700007554.mm.1 | NM_009800.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007554.mm.1 | NM_009800.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007555.mm.1 | TC0700000860.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700007555.mm.1 | NM_009800.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007555.mm.1 | NM_009800.4 | Refseq |
| Mouse Expression Set 430 | 1424379_PM_at | - | - |
| Mouse Expression Set 430 | 1424379_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424379_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424379_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424379_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424379_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 920548 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4920372 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004920372 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2604891 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2604891 | - | - |
| mouse-6 v1.1 (Illumina) | scl32669.8.1_3-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4920372 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32669.8.1_3-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4920372 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1248563 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1248563 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32669.8.1_3-S | - | - |

Car11
Genomic Map