Assay Details
| Gene Symbol: Slc6a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146013.1 | NP_001139485.1 | 12 - 13 | 1951 | 76 | |
| NM_148931.3 | NP_683733.2 | 11 - 12 | 1869 | 76 | ||
| GenBank mRNA | AB118159.1 | - | 12 - 13 | 1898 | 76 | |
| BC138125.1 | - | 11 - 12 | 1872 | 8861144 | 76 | |
| BC138126.1 | - | 11 - 12 | 1872 | 8861145 | 76 | |
| BC157997.1 | - | 12 - 13 | 1951 | 9007308 | 76 | |
| BC172125.1 | - | 11 - 12 | 1812 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a5
Genomic Map