Assay Details
| Gene Symbol: Slc6a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146013.1 | NP_001139485.1 | 9 - 10 | 1583 | 114 | |
| NM_148931.3 | NP_683733.2 | 8 - 9 | 1501 | 114 | ||
| GenBank mRNA | AB118159.1 | - | 9 - 10 | 1530 | 114 | |
| AF411042.3 | - | 9 - 10 | 1637 | 114 | ||
| AY147186.2 | - | 9 - 10 | 1736 | 114 | ||
| BC138125.1 | - | 8 - 9 | 1504 | 8861144 | 114 | |
| BC138126.1 | - | 8 - 9 | 1504 | 8861145 | 114 | |
| BC157997.1 | - | 9 - 10 | 1583 | 9007308 | 114 | |
| BC172125.1 | - | 8 - 9 | 1444 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a5
Genomic Map