Assay Details
| Gene Symbol: Nomo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153057.4 | NP_694697.3 | 19 - 20 | 2387 | 66 | |
| GenBank mRNA | AK030584.1 | - | 19 - 20 | 2354 | 66 | |
| AK035957.1 | - | 19 - 20 | 2359 | 66 | ||
| AK080754.1 | - | 19 - 20 | 2250 | 66 | ||
| AK082495.1 | - | 19 - 20 | 2266 | 66 | ||
| AK082963.1 | - | 19 - 20 | 2246 | 66 | ||
| AK142338.1 | - | 19 - 20 | 2357 | 66 | ||
| AK165643.1 | - | 19 - 20 | 2250 | 66 | ||
| AK166764.1 | - | 4 - 5 | 397 | 66 | ||
| BC022138.1 | - | 18 - 19 | 2103 | 66 | ||
| BC025133.1 | - | 2 - 3 | 266 | 66 | ||
| BC033923.1 | - | 8 - 9 | 971 | 5099899 | 66 | |
| BC072630.1 | - | 19 - 20 | 2333 | 6833516 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nomo1
Genomic Map