Assay Details
| Gene Symbol: Nomo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153057.4 | NP_694697.3 | 11 - 12 | 1330 | 111 | |
| GenBank mRNA | AK030584.1 | - | 11 - 12 | 1297 | 111 | |
| AK035957.1 | - | 11 - 12 | 1300 | 111 | ||
| AK080754.1 | - | 11 - 12 | 1193 | 111 | ||
| AK082495.1 | - | 11 - 12 | 1209 | 111 | ||
| AK082963.1 | - | 11 - 12 | 1189 | 111 | ||
| AK142338.1 | - | 11 - 12 | 1300 | 111 | ||
| AK150107.1 | - | 11 - 12 | 1284 | 111 | ||
| AK165643.1 | - | 11 - 12 | 1193 | 113 | ||
| BC022138.1 | - | 10 - 11 | 1046 | 111 | ||
| BC072630.1 | - | 11 - 12 | 1276 | 6833516 | 111 | |
More Information
Set Membership: |
|

Nomo1
Genomic Map