Assay Details
| Gene Symbol: Gtf2h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291075.1 | NP_001278004.1 | 7 - 8 | 925 | 112 | |
| NM_008186.4 | NP_032212.3 | 6 - 7 | 958 | 112 | ||
| GenBank mRNA | AK005065.1 | - | 6 - 7 | 950 | 112 | |
| BC052837.1 | - | 7 - 8 | 937 | 30049274 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2h1
Genomic Map