Assay Details
| Gene Symbol: Rasa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039103.3 | NP_001034192.1 | 11 - 12 | 1265 | 65 | |
| NM_133914.3 | NP_598675.2 | 11 - 12 | 1265 | 65 | ||
| GenBank mRNA | AK014220.1 | - | 10 - 11 | 1414 | 65 | |
| AK133565.1 | - | 10 - 11 | 1414 | 65 | ||
| AK154335.1 | - | 11 - 12 | 1266 | 65 | ||
| AK155146.1 | - | 11 - 12 | 1266 | 65 | ||
| AK220218.1 | - | 11 - 12 | 1141 | 65 | ||
| AY591339.1 | - | 11 - 12 | 1166 | 65 | ||
| BC027230.1 | - | 10 - 11 | 1386 | 65 | ||
| BC049381.1 | - | 10 - 11 | 1470 | 65 | ||
| BC057460.1 | - | 11 - 12 | 1119 | 4527934 | 65 | |
| DQ317932.1 | - | 11 - 12 | 1149 | 65 | ||
More Information
Set Membership: |
|

Rasa4
Genomic Map