Assay Details
| Gene Symbol: Rasa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039103.3 | NP_001034192.1 | 16 - 17 | 2024 | 63 | |
| NM_133914.3 | NP_598675.2 | 17 - 18 | 2162 | 63 | ||
| GenBank mRNA | AK014220.1 | - | 16 - 17 | 2311 | 63 | |
| AK044762.1 | - | 15 - 16 | 2107 | 63 | ||
| AK154335.1 | - | 17 - 18 | 2163 | 63 | ||
| AK155146.1 | - | 17 - 18 | 2163 | 63 | ||
| AK220218.1 | - | 17 - 18 | 2038 | 63 | ||
| AY591339.1 | - | 17 - 18 | 2063 | 63 | ||
| BC027230.1 | - | 16 - 17 | 2283 | 63 | ||
| BC049381.1 | - | 16 - 17 | 2657 | 63 | ||
| BC057460.1 | - | 17 - 18 | 2016 | 4527934 | 63 | |
| DQ317932.1 | - | 16 - 17 | 1908 | 63 | ||
More Information
Set Membership: |
|

Rasa4
Genomic Map