Assay Details
| Gene Symbol: Rasa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039103.3 | NP_001034192.1 | 2 - 3 | 319 | 61 | |
| NM_133914.3 | NP_598675.2 | 2 - 3 | 319 | 61 | ||
| GenBank mRNA | AK044762.1 | - | 2 - 3 | 147 | 61 | |
| AK133565.1 | - | 2 - 3 | 238 | 61 | ||
| AK154335.1 | - | 2 - 3 | 320 | 61 | ||
| AK155146.1 | - | 2 - 3 | 320 | 61 | ||
| AK220218.1 | - | 2 - 3 | 195 | 61 | ||
| AY591339.1 | - | 2 - 3 | 220 | 61 | ||
| BC027230.1 | - | 2 - 3 | 217 | 61 | ||
| BC049381.1 | - | 2 - 3 | 294 | 61 | ||
| BC057460.1 | - | 2 - 3 | 173 | 4527934 | 61 | |
| BY195753.1 | - | 2 - 3 | 308 | 61 | ||
| DQ317932.1 | - | 2 - 3 | 203 | 61 | ||
More Information
Set Membership: |
|

Rasa4
Genomic Map