Assay Details
| Gene Symbol: Rasa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039103.3 | NP_001034192.1 | 6 - 7 | 707 | 67 | |
| NM_133914.3 | NP_598675.2 | 6 - 7 | 707 | 67 | ||
| GenBank mRNA | AK014220.1 | - | 6 - 7 | 626 | 67 | |
| AK044762.1 | - | 6 - 7 | 535 | 67 | ||
| AK133565.1 | - | 6 - 7 | 626 | 67 | ||
| AK154335.1 | - | 6 - 7 | 708 | 67 | ||
| AK155146.1 | - | 6 - 7 | 708 | 67 | ||
| AK220218.1 | - | 6 - 7 | 583 | 67 | ||
| AY591339.1 | - | 6 - 7 | 608 | 67 | ||
| BC027230.1 | - | 6 - 7 | 598 | 67 | ||
| BC049381.1 | - | 6 - 7 | 682 | 67 | ||
| BC057460.1 | - | 6 - 7 | 561 | 4527934 | 67 | |
| DQ317932.1 | - | 6 - 7 | 591 | 67 | ||
More Information
Set Membership: |
|

Rasa4
Genomic Map