Assay Details
| Gene Symbol: Rasa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039103.3 | NP_001034192.1 | 10 - 11 | 1200 | 74 | |
| NM_133914.3 | NP_598675.2 | 10 - 11 | 1200 | 74 | ||
| GenBank mRNA | AK154335.1 | - | 10 - 11 | 1201 | 74 | |
| AK155146.1 | - | 10 - 11 | 1201 | 74 | ||
| AK220218.1 | - | 10 - 11 | 1076 | 74 | ||
| AY591339.1 | - | 10 - 11 | 1101 | 74 | ||
| BC057460.1 | - | 10 - 11 | 1054 | 4527934 | 74 | |
| DQ317932.1 | - | 10 - 11 | 1084 | 74 | ||
More Information
Set Membership: |
|

Rasa4
Genomic Map