Assay Details
Gene Symbol: Mfsd7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK089227.1 | - | 1 | 1786 | 89 | |
AK138663.1 | - | 1 | 2316 | 89 | ||
AK171069.1 | - | 1 | 1787 | 89 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
