| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500032543.mm.1 |
TC0500003552.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500032543.mm.1 |
NM_017467.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500032543.mm.1 |
NM_017467.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500032546.mm.1 |
TC0500003552.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500032546.mm.1 |
NM_017467.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500032546.mm.1 |
NM_017467.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500032544.mm.1 |
TC0500003552.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500032544.mm.1 |
NM_017467.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500032544.mm.1 |
NM_017467.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500032545.mm.1 |
TC0500003552.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500032545.mm.1 |
NM_017467.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500032545.mm.1 |
NM_017467.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500032547.mm.1 |
TC0500003552.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500032547.mm.1 |
NM_017467.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500032547.mm.1 |
NM_017467.3 |
Refseq |