Assay Details
| Gene Symbol: Zfp316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017467.3 | NP_059495.3 | 3 - 4 | 693 | 148 | |
| GenBank mRNA | BC057078.1 | - | 3 - 4 | 610 | 5698016 | 148 |
| BC060099.1 | - | 1 - 2 | 216 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp316
Genomic Map