Assay Details
| Gene Symbol: Zfp316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017467.3 | NP_059495.3 | 4 - 5 | 783 | 109 | |
| GenBank mRNA | BC057078.1 | - | 4 - 5 | 700 | 5698016 | 109 |
| BC060099.1 | - | 2 - 3 | 306 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp316
Genomic Map